Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PKHD1L1
Entrez Gene ID
93035
Gene Name
polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1
Gene Aliases
PKHDL1
Chromosome Location
Chr.8: 109362477 - 109536809 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177531.4 NP_803875.2 45 - 46 6976 56
XM_011517371.2 XP_011515673.1 45 - 46 6927 56
XM_017013969.1 XP_016869458.1 45 - 46 6927 56
XM_017013970.1 XP_016869459.1 44 - 45 6813 56
XM_017013971.1 XP_016869460.1 45 - 46 6765 56
XM_017013972.1 XP_016869461.1 43 - 44 6624 56
XM_017013973.1 XP_016869462.1 21 - 22 3983 56
XM_017013974.1 XP_016869463.1 16 - 17 3483 56
GenBank mRNA AY219181.1 - 45 - 46 6976 56

View Full Product Details