The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177531.4 | NP_803875.2 | 64 - 65 | 10581 | 82 |
XM_011517371.2 | XP_011515673.1 | 64 - 65 | 10532 | 82 | |
XM_017013969.1 | XP_016869458.1 | 64 - 65 | 10532 | 82 | |
XM_017013970.1 | XP_016869459.1 | 63 - 64 | 10418 | 82 | |
XM_017013971.1 | XP_016869460.1 | 64 - 65 | 10370 | 82 | |
XM_017013972.1 | XP_016869461.1 | 62 - 63 | 10229 | 82 | |
XM_017013973.1 | XP_016869462.1 | 40 - 41 | 7588 | 82 | |
XM_017013974.1 | XP_016869463.1 | 35 - 36 | 7088 | 82 | |
GenBank mRNA | AK131338.1 | - | 10 - 11 | 1263 | 82 |
AY219181.1 | - | 64 - 65 | 10581 | 82 |