Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASUN
Entrez Gene ID
55726
Gene Name
asunder, spermatogenesis regulator
Gene Aliases
C12orf11, GCT1, Mat89Bb, NET48, SPATA30
Chromosome Location
Chr.12: 26905171 - 26938326 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018164.2 NP_060634.2 10 - 11 1599 76
XM_005253422.3 XP_005253479.1 10 - 11 1558 76
XM_011520750.2 XP_011519052.1 10 - 11 1150 76
XM_011520751.2 XP_011519053.1 10 - 11 1576 76
XM_011520753.2 XP_011519055.1 4 - 5 322 76
XM_017019632.1 XP_016875121.1 10 - 11 1132 76
XM_017019633.1 XP_016875122.1 9 - 10 1542 76
XM_017019634.1 XP_016875123.1 9 - 10 1524 76
XM_017019635.1 XP_016875124.1 10 - 11 1132 76
XM_017019637.1 XP_016875126.1 8 - 9 762 76
GenBank mRNA AF274950.1 - 2 - 3 181 76
AK001222.1 - 1 - 2 84 76
AK001492.1 - 10 - 11 1228 76
AK001499.1 - 10 - 11 1264 76
AK225336.1 - 10 - 11 1250 76
AK297948.1 - 8 - 9 939 76
AY211927.1 - 3 - 4 259 76
BC003081.2 - 10 - 11 1288 76
BC008368.1 - 10 - 11 1246 76

View Full Product Details