Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOF
Entrez Gene ID
26509
Gene Name
myoferlin
Gene Aliases
FER1L3
Chromosome Location
Chr.10: 93306429 - 93482504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013451.3 NP_038479.1 21 - 22 2123 91
NM_133337.2 NP_579899.1 20 - 21 2084 91
XM_005269693.4 XP_005269750.1 21 - 22 2302 91
XM_005269694.4 XP_005269751.1 20 - 21 2263 91
XM_011539632.2 XP_011537934.1 22 - 23 2496 91
XM_017016068.1 XP_016871557.1 20 - 21 1883 91
XM_017016069.1 XP_016871558.1 20 - 21 1928 91
XM_017016070.1 XP_016871559.1 21 - 22 2310 91
GenBank mRNA AB033033.2 - 21 - 22 2000 91
AF182316.1 - 21 - 22 2096 91
BC052617.1 - 21 - 22 2088 91

View Full Product Details