The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013451.3 | NP_038479.1 | 33 - 34 | 3779 | 129 |
NM_133337.2 | NP_579899.1 | 32 - 33 | 3740 | 129 | |
XM_005269693.4 | XP_005269750.1 | 33 - 34 | 3958 | 129 | |
XM_005269694.4 | XP_005269751.1 | 32 - 33 | 3919 | 129 | |
XM_011539632.2 | XP_011537934.1 | 34 - 35 | 4152 | 129 | |
XM_017016068.1 | XP_016871557.1 | 32 - 33 | 3539 | 129 | |
XM_017016069.1 | XP_016871558.1 | 32 - 33 | 3584 | 129 | |
XM_017016070.1 | XP_016871559.1 | 33 - 34 | 3966 | 129 | |
GenBank mRNA | AB033033.2 | - | 33 - 34 | 3656 | 129 |
AF182316.1 | - | 33 - 34 | 3752 | 129 | |
AF207990.1 | - | 32 - 33 | 3617 | 129 | |
AL096713.1 | - | 13 - 14 | 1796 | 129 |