Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYOF
Entrez Gene ID
26509
Gene Name
myoferlin
Gene Aliases
FER1L3
Chromosome Location
Chr.10: 93306429 - 93482504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013451.3 NP_038479.1 2 - 3 266 119
NM_133337.2 NP_579899.1 2 - 3 266 119
XM_005269693.4 XP_005269750.1 2 - 3 445 119
XM_005269694.4 XP_005269751.1 2 - 3 445 119
XM_017016070.1 XP_016871559.1 2 - 3 453 119
GenBank mRNA AB033033.2 - 2 - 3 143 119
AF182316.1 - 2 - 3 239 119
AF207990.1 - 2 - 3 143 119
BC052617.1 - 2 - 3 231 119

View Full Product Details