The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005236.2 | NP_005227.1 | 7 - 8 | 1218 | 91 |
XM_011522424.2 | XP_011520726.1 | 8 - 9 | 1375 | 91 | |
XM_011522427.1 | XP_011520729.1 | 2 - 3 | 355 | 91 | |
XM_017023043.1 | XP_016878532.1 | 8 - 9 | 1332 | 91 | |
GenBank mRNA | AK289726.1 | - | 7 - 8 | 1217 | 91 |
AK301930.1 | - | 2 - 3 | 487 | 91 | |
AK308341.1 | - | 7 - 8 | 1184 | 91 | |
U64315.1 | - | 7 - 8 | 1191 | 91 |