Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GPS1
- Entrez Gene ID
-
2873
- Gene Name
- G protein pathway suppressor 1
- Gene Aliases
-
COPS1, CSN1, SGN1
- Chromosome Location
-
Chr.17: 82051477 - 82057470 on Build GRCh38
View Full Product Details