Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGT7
Entrez Gene ID
2686
Gene Name
gamma-glutamyltransferase 7
Gene Aliases
D20S101, GGT4, GGTL3, GGTL5, dJ18C9.2
Chromosome Location
Chr.20: 34844720 - 34872858 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178026.2 NP_821158.2 9 - 10 1279 62
XM_011528782.2 XP_011527084.1 7 - 8 758 62
XM_011528783.2 XP_011527085.1 4 - 5 527 62
XM_017027809.1 XP_016883298.1 9 - 10 1278 62
XM_017027810.1 XP_016883299.1 9 - 10 1278 62
XM_017027811.1 XP_016883300.1 9 - 10 1278 62
GenBank mRNA AK090413.1 - 2 - 3 3247 62
BC033745.1 - 9 - 10 1279 62
BC040030.1 - 10 - 11 1294 62
BC101371.1 - 9 - 10 1268 62
BC101372.1 - 9 - 10 1275 62
BC141843.1 - 9 - 10 1268 62

View Full Product Details