Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLA2R1
Entrez Gene ID
22925
Gene Name
phospholipase A2 receptor 1
Gene Aliases
CLEC13C, PLA2-R, PLA2G1R, PLA2IR, PLA2R
Chromosome Location
Chr.2: 159928384 - 160062902 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007267.2 NP_001007268.1 6 - 7 1305 107
NM_001195641.1 NP_001182570.1 6 - 7 1305 107
NM_007366.4 NP_031392.3 6 - 7 1305 107
XM_005246392.3 XP_005246449.1 6 - 7 1193 107
XM_011510820.2 XP_011509122.1 6 - 7 1193 107
XM_017003598.1 XP_016859087.1 6 - 7 1193 107
XM_017003599.1 XP_016859088.1 6 - 7 1193 107
XM_017003601.1 XP_016859090.1 6 - 7 1592 107
GenBank mRNA BC140823.1 - 6 - 7 1115 107
BC144631.1 - 6 - 7 1115 107
U17033.1 - 6 - 7 1305 107
U17034.1 - 6 - 7 1305 107

View Full Product Details