The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133445.2 | NP_597702.2 | 2 - 3 | 1910 | 81 |
XM_011518211.2 | XP_011516513.1 | 2 - 3 | 1334 | 81 | |
GenBank mRNA | AB075853.1 | - | 2 - 3 | 1910 | 81 |
AF416558.1 | - | 2 - 3 | 1322 | 81 | |
AJ416950.2 | - | 2 - 3 | 1324 | 81 | |
AK127342.1 | - | 2 - 3 | 877 | 81 | |
BC132866.1 | - | 2 - 3 | 1736 | 81 | |
BC132868.1 | - | 2 - 3 | 1736 | 81 | |
BC167432.1 | - | 2 - 3 | 1910 | 81 |