Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UTP20
Entrez Gene ID
27340
Gene Name
UTP20, small subunit processome component
Gene Aliases
1A6/DRIM, DRIM
Chromosome Location
Chr.12: 101280127 - 101386619 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014503.2 NP_055318.2 33 - 34 4398 150
GenBank mRNA AJ006778.1 - 33 - 34 4386 150

View Full Product Details