The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014503.2 | NP_055318.2 | 36 - 37 | 4757 | 112 |
GenBank mRNA | AJ006778.1 | - | 36 - 37 | 4745 | 112 |