The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014503.2 | NP_055318.2 | 52 - 53 | 7031 | 72 |
GenBank mRNA | AF072718.1 | - | 10 - 11 | 1230 | 72 |
AJ006778.1 | - | 52 - 53 | 7019 | 72 | |
AK000333.1 | - | 7 - 8 | 814 | 72 | |
AK297739.1 | - | 3 - 4 | 232 | 72 |