The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291476.1 | NP_001278405.1 | 11 - 12 | 1509 | 103 |
NM_007010.4 | NP_008941.3 | 10 - 11 | 1394 | 103 | |
XM_011524232.2 | XP_011522534.1 | 10 - 11 | 1378 | 103 | |
XM_011524233.2 | XP_011522535.1 | 8 - 9 | 1250 | 103 | |
GenBank mRNA | AB208938.1 | - | 10 - 11 | 1348 | 103 |
AF077033.1 | - | 10 - 11 | 1361 | 103 | |
AJ010840.1 | - | 7 - 8 | 813 | 103 | |
AK001245.1 | - | 7 - 8 | 903 | 103 | |
AK001652.1 | - | 11 - 12 | 1490 | 103 | |
AK027425.1 | - | 7 - 8 | 854 | 103 | |
BC041785.1 | - | 10 - 11 | 1376 | 103 |