The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004907.2 | NP_004898.2 | 1 - 2 | 86 | 71 |
GenBank mRNA | BC072432.1 | - | 1 - 2 | 86 | 71 |