The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293725.1 | NP_001280654.1 | 3 - 4 | 649 | 65 |
NM_005640.2 | NP_005631.1 | 3 - 4 | 649 | 65 | |
NR_121653.1 | 3 - 4 | 649 | 65 | ||
XM_005258339.3 | XP_005258396.1 | 3 - 4 | 1041 | 65 | |
XM_011526153.2 | XP_011524455.1 | 3 - 4 | 1041 | 65 | |
XM_017025932.1 | XP_016881421.1 | 3 - 4 | 1041 | 65 | |
XM_017025933.1 | XP_016881422.1 | 3 - 4 | 273 | 65 | |
GenBank mRNA | AB234096.1 | - | 3 - 4 | 1046 | 65 |
AK302593.1 | - | 2 - 3 | 138 | 65 | |
AY888048.1 | - | 3 - 4 | 649 | 65 | |
AY888049.1 | - | 3 - 4 | 649 | 65 | |
Y09321.1 | - | 3 - 4 | 400 | 65 |