The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293725.1 | NP_001280654.1 | 4 - 5 | 810 | 117 |
NM_005640.2 | NP_005631.1 | 4 - 5 | 810 | 117 | |
NR_121653.1 | 4 - 5 | 810 | 117 | ||
XM_005258339.3 | XP_005258396.1 | 4 - 5 | 1202 | 117 | |
XM_011526153.2 | XP_011524455.1 | 4 - 5 | 1202 | 117 | |
XM_017025932.1 | XP_016881421.1 | 4 - 5 | 1202 | 117 | |
XM_017025933.1 | XP_016881422.1 | 4 - 5 | 434 | 117 | |
GenBank mRNA | AB234096.1 | - | 4 - 5 | 1207 | 117 |
AK302593.1 | - | 3 - 4 | 299 | 117 | |
AY888048.1 | - | 4 - 5 | 810 | 117 | |
AY888049.1 | - | 4 - 5 | 810 | 117 | |
Y09321.1 | - | 4 - 5 | 561 | 117 |