The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293725.1 | NP_001280654.1 | 9 - 10 | 1898 | 101 |
NM_005640.2 | NP_005631.1 | 9 - 10 | 1883 | 101 | |
XM_017025933.1 | XP_016881422.1 | 9 - 10 | 1507 | 101 | |
GenBank mRNA | AB234096.1 | - | 9 - 10 | 2295 | 101 |
AK302593.1 | - | 8 - 9 | 1372 | 101 | |
AY888048.1 | - | 9 - 10 | 1883 | 101 | |
Y09321.1 | - | 9 - 10 | 1649 | 101 |