Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FNBP1
Entrez Gene ID
23048
Gene Name
formin binding protein 1
Gene Aliases
FBP17
Chromosome Location
Chr.9: 129887187 - 130053846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015033.2 NP_055848.1 6 - 7 738 82
XM_005251815.2 XP_005251872.1 6 - 7 711 82
XM_005251820.2 XP_005251877.1 6 - 7 711 82
XM_005251821.2 XP_005251878.1 6 - 7 711 82
XM_005251822.2 XP_005251879.1 6 - 7 711 82
XM_005251823.2 XP_005251880.1 6 - 7 711 82
XM_005251824.2 XP_005251881.1 6 - 7 711 82
XM_005251825.2 XP_005251882.1 6 - 7 711 82
XM_005251826.2 XP_005251883.1 6 - 7 711 82
XM_005251827.2 XP_005251884.1 6 - 7 711 82
XM_005251828.2 XP_005251885.1 6 - 7 711 82
XM_005251829.2 XP_005251886.1 6 - 7 711 82
XM_005251830.2 XP_005251887.1 6 - 7 711 82
XM_005251831.2 XP_005251888.1 6 - 7 711 82
XM_005251832.2 XP_005251889.1 6 - 7 711 82
XM_005251833.2 XP_005251890.1 6 - 7 711 82
XM_005251834.2 XP_005251891.1 6 - 7 711 82
XM_006717016.2 XP_006717079.1 6 - 7 711 82
XM_011518399.2 XP_011516701.1 6 - 7 711 82
XM_011518400.2 XP_011516702.1 6 - 7 711 82
XM_011518401.1 XP_011516703.1 6 - 7 711 82
XM_011518402.1 XP_011516704.1 6 - 7 711 82
XM_017014487.1 XP_016869976.1 6 - 7 711 82
XM_017014488.1 XP_016869977.1 6 - 7 711 82
XM_017014489.1 XP_016869978.1 6 - 7 711 82
XM_017014490.1 XP_016869979.1 6 - 7 711 82
XM_017014491.1 XP_016869980.1 6 - 7 711 82
XM_017014492.1 XP_016869981.1 6 - 7 997 82
XM_017014493.1 XP_016869982.1 6 - 7 711 82
XM_017014494.1 XP_016869983.1 6 - 7 711 82
GenBank mRNA AB011126.1 - 6 - 7 692 82
AF265550.1 - 6 - 7 707 82
AK000975.1 - 5 - 6 415 82
AK001616.1 - 5 - 6 406 82
AK023681.1 - 6 - 7 711 82
BC062463.1 - 6 - 7 653 82
BC101755.1 - 6 - 7 580 82
BC143513.1 - 6 - 7 580 82
BC143514.1 - 6 - 7 580 82
BC143515.1 - 6 - 7 580 82

View Full Product Details