Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YWHAZ
Entrez Gene ID
7534
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein zeta
Gene Aliases
14-3-3-zeta, HEL-S-3, HEL-S-93, HEL4, KCIP-1, YWHAD
Chromosome Location
Chr.8: 100918576 - 100954068 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135699.1 NP_001129171.1 2 - 3 447 120
NM_001135700.1 NP_001129172.1 2 - 3 401 120
NM_001135701.1 NP_001129173.1 2 - 3 450 120
NM_001135702.1 NP_001129174.1 2 - 3 469 120
NM_003406.3 NP_003397.1 2 - 3 430 120
NM_145690.2 NP_663723.1 2 - 3 504 120
XM_005251061.3 XP_005251118.1 4 - 5 827 120
XM_005251063.3 XP_005251120.1 2 - 3 599 120
XM_017013810.1 XP_016869299.1 3 - 4 616 120
XM_017013811.1 XP_016869300.1 4 - 5 812 120
GenBank mRNA AK289945.1 - 2 - 3 340 120
AK307506.1 - 3 - 4 307 120
AK315867.1 - 2 - 3 218 120
BC003623.2 - 2 - 3 397 120
BC013265.1 - 2 - 3 395 120
BC051814.1 - 2 - 3 518 120
BC063824.1 - 2 - 3 369 120
BC068456.1 - 2 - 3 418 120
BC072426.1 - 2 - 3 399 120
BC073141.1 - 2 - 3 353 120
BC083508.1 - 2 - 3 397 120
BC099904.1 - 2 - 3 419 120
BC101483.1 - 2 - 3 352 120
BC108281.1 - 2 - 3 407 120
BC111951.1 - 2 - 3 352 120
CN276627.1 - 2 - 3 395 120
CR996247.1 - 2 - 3 401 120
DC298291.1 - 2 - 3 430 120
EU794582.1 - 2 - 3 398 120
EU794583.1 - 2 - 3 418 120
GQ472215.1 - 2 - 3 418 120
M86400.1 - 2 - 3 376 120
U28964.1 - 2 - 3 418 120

View Full Product Details