Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYH2
Entrez Gene ID
4620
Gene Name
myosin heavy chain 2
Gene Aliases
IBM3, MYH2A, MYHSA2, MYHas8, MYPOP, MyHC-2A, MyHC-IIa
Chromosome Location
Chr.17: 10521148 - 10549700 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100112.1 NP_001093582.1 13 - 14 1398 59
NM_017534.5 NP_060004.3 13 - 14 1398 59
GenBank mRNA AF111784.1 - 11 - 12 1270 59
AK299440.1 - 13 - 14 1209 59
BC093082.1 - 12 - 13 1343 59
BC126409.1 - 12 - 13 1341 59
BX510904.2 - 13 - 14 1361 59

View Full Product Details