The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011527471.2 | XP_011525773.1 | 32 - 33 | 3966 | 97 |
XM_011527472.2 | XP_011525774.1 | 32 - 33 | 3951 | 97 | |
XM_011527474.2 | XP_011525776.1 | 31 - 32 | 3846 | 97 | |
XM_017027459.1 | XP_016882948.1 | 22 - 23 | 2949 | 97 | |
GenBank mRNA | AK023134.1 | - | 6 - 7 | 761 | 97 |