Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCAF4
Entrez Gene ID
57466
Gene Name
SR-related CTD associated factor 4
Gene Aliases
SFRS15, SRA4
Chromosome Location
Chr.21: 31671000 - 31732118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145444.1 NP_001138916.1 9 - 10 1613 119
NM_001145445.1 NP_001138917.1 10 - 11 1658 119
NM_020706.2 NP_065757.1 10 - 11 1658 119
XM_005261017.2 XP_005261074.1 9 - 10 1462 119
XM_017028415.1 XP_016883904.1 10 - 11 1616 119
XM_017028416.1 XP_016883905.1 10 - 11 1501 119
XM_017028417.1 XP_016883906.1 9 - 10 1462 119
GenBank mRNA AB032998.1 - 4 - 5 646 119
AF023142.1 - 10 - 11 1262 119
AK025748.1 - 5 - 6 677 119
AK308406.1 - 9 - 10 1464 119
AL117417.1 - 5 - 6 666 119
AL834304.1 - 10 - 11 1438 119
BC014921.1 - 10 - 11 1635 119
BC043353.1 - 10 - 11 1615 119
BC052286.1 - 10 - 11 1489 119
BC064990.1 - 10 - 11 1489 119

View Full Product Details