The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012294.3 | NP_036426.3 | 7 - 8 | 651 | 148 |
XM_005249914.2 | XP_005249971.1 | 7 - 8 | 870 | 148 | |
XM_005249916.1 | XP_005249973.1 | 7 - 8 | 814 | 148 | |
XM_017012837.1 | XP_016868326.1 | 5 - 6 | 555 | 148 | |
GenBank mRNA | AL833195.1 | - | 7 - 8 | 630 | 148 |