The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199861.1 | NP_001186790.1 | 1 - 2 | 494 | 96 |
XM_005263514.2 | XP_005263571.1 | 1 - 2 | 191 | 96 | |
XM_011542321.2 | XP_011540623.1 | 1 - 2 | 166 | 96 | |
XM_017002619.1 | XP_016858108.1 | 1 - 2 | 72 | 96 | |
GenBank mRNA | AK289819.1 | - | 1 - 2 | 100 | 96 |
AK290044.1 | - | 1 - 2 | 72 | 96 | |
AK312395.1 | - | 1 - 2 | 72 | 96 | |
AK315811.1 | - | 1 - 2 | 86 | 96 |