Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYBB
Entrez Gene ID
1536
Gene Name
cytochrome b-245 beta chain
Gene Aliases
AMCBX2, CGD, GP91-1, GP91-PHOX, GP91PHOX, IMD34, NOX2, p91-PHOX
Chromosome Location
Chr.X: 37780017 - 37813461 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000397.3 NP_000388.2 1 - 2 102 80
GenBank mRNA AK289753.1 - 1 - 2 87 80
AK301476.1 - 1 - 2 55 80
AK304033.1 - 1 - 2 62 80
BC032720.1 - 1 - 2 88 80
DA995835.1 - 1 - 2 102 80
KU178009.1 - 1 - 2 41 80
KU178010.1 - 1 - 2 41 80

View Full Product Details