Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNAJB11
Entrez Gene ID
51726
Gene Name
DnaJ heat shock protein family (Hsp40) member B11
Gene Aliases
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
Chromosome Location
Chr.3: 186570676 - 186585800 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016306.5 NP_057390.1 5 - 6 814 82
GenBank mRNA AB028859.2 - 5 - 6 757 82
AF228505.1 - 5 - 6 683 82
AF277317.1 - 5 - 6 774 82
AJ250137.1 - 5 - 6 612 82
AK075300.1 - 5 - 6 771 82
AK075430.1 - 5 - 6 790 82
AK124289.1 - 1 - 2 3210 82
AK301292.1 - 3 - 4 473 82
AY359043.1 - 5 - 6 750 82
BC001144.1 - 5 - 6 752 82
BC046500.1 - 6 - 7 1521 82
BT007063.1 - 5 - 6 592 82
CR457096.1 - 5 - 6 592 82

View Full Product Details