The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144822.1 | NP_001138294.1 | 1 - 2 | 188 | 157 |
NM_001779.2 | NP_001770.1 | 1 - 2 | 188 | 157 | |
NR_026665.1 | 1 - 2 | 188 | 157 | ||
XM_017002869.1 | XP_016858358.1 | 1 - 2 | 188 | 157 | |
GenBank mRNA | AK291980.1 | - | 1 - 2 | 121 | 157 |
BC005930.1 | - | 1 - 2 | 134 | 157 | |
X06296.1 | - | 1 - 2 | 79 | 157 | |
Y00636.1 | - | 1 - 2 | 76 | 157 |