Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DNAJB11
- Entrez Gene ID
-
51726
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B11
- Gene Aliases
-
ABBP-2, ABBP2, DJ9, Dj-9, EDJ, ERdj3, ERj3, ERj3p, PRO1080, UNQ537
- Chromosome Location
-
Chr.3: 186570676 - 186585800 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC046500.1 |
- |
1 - 2 |
193 |
97 |
View Full Product Details