Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD86
Entrez Gene ID
942
Gene Name
CD86 molecule
Gene Aliases
B7-2, B7.2, B70, CD28LG2, LAB72
Chromosome Location
Chr.3: 122055362 - 122121143 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206924.1 NP_001193853.1 5 - 6 679 104
NM_001206925.1 NP_001193854.1 5 - 6 965 104
NM_006889.4 NP_008820.3 6 - 7 1070 104
NM_175862.4 NP_787058.4 6 - 7 1015 104
NM_176892.1 NP_795711.1 5 - 6 926 104
GenBank mRNA AK291497.1 - 6 - 7 923 104
AK294663.1 - 5 - 6 675 104
AK301237.1 - 5 - 6 961 104
AK313103.1 - 6 - 7 924 104
AK316203.1 - 5 - 6 961 104
BC040261.1 - 6 - 7 1003 104
CR541844.1 - 5 - 6 869 104
L25259.1 - 6 - 7 993 104
U04343.1 - 6 - 7 1016 104

View Full Product Details