Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FNIP2
Entrez Gene ID
57600
Gene Name
folliculin interacting protein 2
Gene Aliases
FNIPL, MAPO1
Chromosome Location
Chr.4: 158768995 - 158908050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323916.1 NP_001310845.1 4 - 5 711 128
NM_020840.2 NP_065891.1 4 - 5 706 128
XM_005263156.1 XP_005263213.1 4 - 5 706 128
XM_005263158.1 XP_005263215.1 4 - 5 506 128
XM_005263160.3 XP_005263217.1 4 - 5 706 128
XM_011532154.2 XP_011530456.1 4 - 5 1486 128
XM_017008487.1 XP_016863976.1 4 - 5 706 128
GenBank mRNA AB040883.1 - 4 - 5 563 128
AL713802.1 - 4 - 5 507 128
BC016638.1 - 4 - 5 711 128

View Full Product Details