Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CD58
- Entrez Gene ID
-
965
- Gene Name
- CD58 molecule
- Gene Aliases
-
LFA-3, LFA3, ag3
- Chromosome Location
-
Chr.1: 116487140 - 116571093 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
D28586.1 |
- |
1 - 2 |
283 |
110 |
View Full Product Details