The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282290.1 | NP_001269219.1 | 10 - 11 | 1865 | 81 |
NM_199282.2 | NP_954976.1 | 10 - 11 | 1273 | 81 | |
XM_005257128.3 | XP_005257185.1 | 9 - 10 | 1322 | 81 | |
XM_006721745.2 | XP_006721808.1 | 13 - 14 | 2247 | 81 | |
XM_006721747.2 | XP_006721810.1 | 12 - 13 | 2181 | 81 | |
XM_017024312.1 | XP_016879801.1 | 10 - 11 | 1386 | 81 | |
GenBank mRNA | AK125535.1 | - | 10 - 11 | 1258 | 81 |
BC101388.1 | - | 9 - 10 | 1041 | 81 | |
BC101389.1 | - | 9 - 10 | 1041 | 81 | |
BC101390.1 | - | 9 - 10 | 1041 | 81 | |
BC101391.2 | - | 8 - 9 | 960 | 81 |