Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CROCCP3
Entrez Gene ID
114819
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 3
Gene Aliases
CROCCL2, dJ798A10.2
Chromosome Location
Chr.1: 16467436 - 16492701 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_023386.1 1 - 2 721 87
GenBank mRNA AB067509.1 - 1 - 2 720 87


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream NECAP endocytosis associated 2 NECAP2 -31.1kb Anti-sense Chr. - 16440672 - 16460090 on Build GRCh38
Downstream RNA, U1 small nuclear 1 RNU1-1 HSD1, HU1-1, RNU1, RNU1A, RNU1A3, RNU1G4, Rnu1a1, U1, U1A1 21.4kb Sense Chr. - 16514122 - 16514285 on Build GRCh38

View Full Product Details