Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
Chromosome Location
Chr.18: 23992773 - 24135952 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005258198.4 XP_005258255.1 16 - 16 101
XM_011525814.2 XP_011524116.1 13 - 13 101
XM_017025550.1 XP_016881039.1 16 - 16 101


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI871745.1 - 1 Chr. 18 - 24135313 - 24135854 [+] on Build GRCh38 101
GenBankmRNA HY218866.1 - 1 191 Chr. 18 - 24135547 - 24135969 [-] on Build GRCh38 101
GenBankmRNA AV656442.1 - 2 Chr. 18 - 24135499 - 24135919 [+] on Build GRCh38 101
GenBankmRNA AF363068.1 - 51 Chr. - - [-] on Build 101
GenBankmRNA CA774026.1 - 1 190 Chr. 18 - 24135430 - 24135968 [-] on Build GRCh38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream TTC39C antisense RNA 1 TTC39C-AS1 -119.9kb Anti-sense Chr. - 23994213 - 24015460 on Build GRCh38
Downstream calcium binding tyrosine phosphorylation regulated CABYR CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2 3.1kb Sense Chr. - 24138956 - 24161600 on Build GRCh38

View Full Product Details