Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CD58
- Entrez Gene ID
-
965
- Gene Name
- CD58 molecule
- Gene Aliases
-
LFA-3, LFA3, ag3
- Chromosome Location
-
Chr.1: 116487140 - 116571093 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_026665.1 |
|
4 - 5 |
819 |
124 |
GenBank mRNA |
CD365881.1 |
- |
2 - 3 |
|
124 |
X06296.1 |
- |
4 - 5 |
710 |
124 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105378922 |
LOC105378922 |
|
-14.1kb
|
Sense |
Chr. - 116493016 - 116505088 on Build GRCh38 |
Downstream |
microRNA 548ac |
MIR548AC |
|
38.0kb
|
Sense |
Chr. - 116560024 - 116560111 on Build GRCh38 |
View Full Product Details