Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CK817666.1 |
- |
1 |
|
|
Chr. 2 - 32032251 - 32032855 [+] on Build GRCh38 |
73 |
GenBankmRNA |
AK127337.1 |
- |
1 |
3452 |
|
Chr. 2 - 32032057 - 32035967 [-] on Build GRCh38 |
73 |
GenBankmRNA |
BG655497.1 |
- |
1 |
|
|
Chr. 2 - 32032258 - 32032739 [+] on Build GRCh38 |
73 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
dpy-30, histone methyltransferase complex regulatory subunit |
DPY30 |
Cps25, HDPY-30, Saf19 |
|
Sense |
Chr. - 32011649 - 32039840 on Build GRCh38 |
Downstream |
spastin |
SPAST |
ADPSP, FSP2, SPG4 |
27.6kb
|
Anti-sense |
Chr. - 32063551 - 32157637 on Build GRCh38 |
View Full Product Details