Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EIF5AL1
Entrez Gene ID
143244
Gene Name
eukaryotic translation initiation factor 5A-like 1
Gene Aliases
EIF5AP1, bA342M3.3
Chromosome Location
Chr.10: 79512601 - 79516440 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099692.1 NP_001093162.1 1 - 1 3451 97
GenBank mRNA BC070048.1 - 1 - 1 3452 97

View Full Product Details