The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002954.5 | NP_002945.1 | 1 - 2 | 304 | 99 |
GenBank mRNA | BM907473.1 | - | 5 - 6 | 99 | |
CR746519.1 | - | 1 - 2 | 315 | 99 |