Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC103091866
Entrez Gene ID
103091866
Gene Name
uncharacterized LOC103091866
Gene Aliases
-
Chromosome Location
Chr.1: 145168405 - 145216071 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_111957.1 3 - 4 733 67
NR_111958.1 2 - 3 678 67
GenBank mRNA AK307112.1 - 2 - 3 457 67
AK309334.1 - 3 - 4 476 67

Gene Symbol
LINC00869
Entrez Gene ID
57234
Gene Name
long intergenic non-protein coding RNA 869
Gene Aliases
FAM91A2
Chromosome Location
Chr.1: 149607012 - 149679523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA DA711671.1 - 3 - 4 512 67


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA460367.1 - 3 - 4 476 Chr. 1 - 145168994 - 145215814 [-] on Build GRCh38 67
GenBankmRNA CK000370.1 - 2 - 3 597 Chr. 1 - 145168885 - 145215984 [-] on Build GRCh38 67
GenBankmRNA DA368492.1 - 2 - 3 425 Chr. 1 - 145168941 - 145215814 [-] on Build GRCh38 67
GenBankmRNA DR157396.1 - 2 - 3 355 Chr. 1 - 145168655 - 145215744 [-] on Build GRCh38 67
GenBankmRNA DA726693.1 - 2 - 3 457 Chr. 1 - 145169020 - 145215850 [-] on Build GRCh38 67
GenBankmRNA DB278681.1 - 2 - 3 458 Chr. 1 - 145169017 - 145215850 [-] on Build GRCh38 67
GenBankmRNA DA368554.1 - 2 - 3 421 Chr. 1 - 145168917 - 145215814 [-] on Build GRCh38 67
GenBankmRNA DA221467.1 - 2 - 3 457 Chr. 1 - 145169044 - 145215850 [-] on Build GRCh38 67


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream family with sequence similarity 72 member D FAM72D GCUD2 -56.2kb Anti-sense Chr. - 145093805 - 145112696 on Build GRCh38
Downstream uncharacterized LOC100996740 LOC100996740 43.6kb Anti-sense Chr. - 145214738 - 145216546 on Build GRCh38

View Full Product Details