Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LOC101929595
- Entrez Gene ID
-
101929595
- Gene Name
- uncharacterized LOC101929595
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 107863479 - 107932119 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_125929.1 |
|
4 - 5 |
991 |
104 |
GenBank mRNA |
AK123292.1 |
- |
4 - 5 |
991 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AI806251.1 |
- |
3 - 4 |
404 |
|
Chr. 4 - 107873329 - 107893185 [-] on Build GRCh38 |
104 |
GenBankmRNA |
DB034690.1 |
- |
3 - 4 |
394 |
|
Chr. 4 - 107867572 - 107893186 [-] on Build GRCh38 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sphingomyelin synthase 2 |
SGMS2 |
SMS2 |
|
Anti-sense |
Chr. - 107823645 - 107915047 on Build GRCh38 |
Downstream |
cytochrome P450 family 2 subfamily U member 1 |
CYP2U1 |
P450TEC, SPG49, SPG56 |
57.5kb
|
Anti-sense |
Chr. - 107931549 - 107953461 on Build GRCh38 |
View Full Product Details