Gene Transcripts

Gene Symbol
Hpdl
Entrez Gene ID
242642
Gene Name
4-hydroxyphenylpyruvate dioxygenase-like
Gene Aliases
A830048M07Rik, BC034099, Gloxd1
Chromosome Location
Chr. 4: 116819907 - 116821508 on Build GRCm38
UniGene ID
Mm.279897
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146256.3 NP_666368.1 1 798
GenBank mRNA AK043902.1 - 1 779
AK049217.1 - 1 778
AK078929.1 - 1 799
BC034099.1 - 1 613 4489103

View Full Product Details