Gene Transcripts

Gene Symbol
Vps37b
Entrez Gene ID
330192
Gene Name
vacuolar protein sorting 37B (yeast)
Gene Aliases
2300007F24Rik, AI415429, BC026744
Chromosome Location
Chr. 5: 124004641 - 124032260 on Build GRCm38
UniGene ID
Mm.4079
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177876.4 NP_808544.1 3 367
GenBank mRNA AK082658.1 - 3 328
AK149893.1 - 3 334
AK166359.1 - 3 348
AK172114.1 - 3 328
AK172337.1 - 3 328
BC026744.1 - 3 368 4163734

View Full Product Details