Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_148934.2 | NP_683736.1 | 1 | 195 | |
GenBank mRNA | AF303106.1 | - | 1 | 195 | |
AK153467.1 | - | 1 | 122 | ||
BC138515.1 | - | 1 | 236 | 8861535 | |
BC138516.1 | - | 1 | 236 | 8861536 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK077345.1 | - | 615 | Chr. - - [-] |