Product Details
- SNP ID
-
rs6261499
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:50470340 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACAAAGTTTATTTGGTCATTAATG[A/G]GCACTGTGTTTCTAATTTGCTACTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Prkd1
PubMed Links
Gene Details
- Gene
- Prkd1
- Gene Name
- protein kinase D1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008858.3 |
|
Intron |
|
|
NP_032884.2 |
View Full Product Details