Product Details

SNP ID
rs13481895
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:77985933 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTATTGTCCTCTTTGTTTTCGGAA[A/G]GGACTGATAGACACTGCTGTGTGCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Fam172a PubMed Links

Gene Details

Gene
Fam172a
Gene Name
family with sequence similarity 172, member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163419.1 Intron NP_001156891.1
NM_001163420.1 Intron NP_001156892.1
NM_138312.1 Intron NP_612185.1

View Full Product Details