Product Details
- SNP ID
-
rs13481828
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:58051262 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTGAAGGCGAGGAGGAGCTGGAGG[A/G]CAGAGGAGAAAAAGACAGACATCCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Klhl3
PubMed Links
Gene Details
- Gene
- Klhl3
- Gene Name
- kelch-like 3
View Full Product Details