Product Details

SNP ID
rs3677220
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:78017908 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCTGATATTTTTGATCTCCCCATT[T/A]TAAATTTCCATACACTAAGCAGCAA
Phenotype
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
Fam172a PubMed Links

Gene Details

Gene
Fam172a
Gene Name
family with sequence similarity 172, member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163419.1 Intron NP_001156891.1
NM_001163420.1 Intron NP_001156892.1
NM_138312.1 Intron NP_612185.1
Gene
Pou5f2
Gene Name
POU domain class 5, transcription factor 2
There are no transcripts associated with this gene.

View Full Product Details