Product Details

SNP ID
rs13483700
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.19:60757991 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AATGCAATAACTCGAGTTCCAAAAG[A/G]TTCTACTCTGGGCTGTTTGCCTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Eif3a PubMed Links

Gene Details

Gene
Eif3a
Gene Name
eukaryotic translation initiation factor 3, subunit A
There are no transcripts associated with this gene.

Gene
Nanos1
Gene Name
nanos homolog 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178421.3 2005 UTR 3 NP_848508.2

View Full Product Details